This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06438513_m1
See other PDHA1 GE Assays ›
Gene Symbol:
PDHA1
Gene Name:
pyruvate dehydrogenase E1 subunit alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.X: 15699924 - 15718908 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06438513_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDHA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003360244.4 XP_003360292.2 698 82
GenBank mRNA AK239558.1 - 4 - 5 513 82
AK344901.1 - 4 - 5 522 82
AK394526.1 - 4 - 5 573 82
X52990.1 - 413 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
acetyl-CoA biosynthetic process from pyruvate
tricarboxylic acid cycle
pyruvate dehydrogenase (acetyl-transferring) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0