This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06438739_s1
See other UBE2H GE Assays ›
Gene Symbol:
UBE2H
Gene Name:
ubiquitin conjugating enzyme E2 H
Gene Aliases:
-
Chromosome Location:
Chr.18: 18819612 - 18932909 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06438739_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003360138.5 XP_003360186.3 3032 71
XM_021078794.1 XP_020934453.1 2739 71
XM_021078795.1 XP_020934454.1 2961 71
XM_021078796.1 XP_020934455.1 2941 71
XM_021078797.1 XP_020934456.1 5696 71
XM_021078798.1 XP_020934457.1 2588 71
XM_021078799.1 XP_020934458.1 2689 71
GenBank mRNA AK394218.1 - 1 617 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0