This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06826364_m1
See other ISLR2 GE Assays ›
Gene Symbol:
ISLR2
Gene Name:
immunoglobulin superfamily containing leucine rich repeat 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 59385021 - 59393185 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06826364_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ISLR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005666160.3 XP_005666217.1 626 91
XM_005666161.3 XP_005666218.1 414 91
XM_005666162.3 XP_005666219.1 349 91
XM_005666164.3 XP_005666221.1 423 91
XM_013978091.2 XP_013833545.1 275 91
XM_013978093.2 XP_013833547.1 1256 91
XM_013978094.2 XP_013833548.1 481 91

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005666160.3, XM_005666161.3, XM_005666162.3, XM_005666164.3, XM_013978093.2, XM_013978094.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0