This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06830143_g1
See other GPAT2 GE Assays ›
Gene Symbol:
GPAT2
Gene Name:
glycerol-3-phosphate acyltransferase 2, mitochondrial
Gene Aliases:
-
Chromosome Location:
Chr.3: 45843666 - 45857406 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06830143_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013995842.2 XP_013851296.1 1782 121
XM_021087050.1 XP_020942709.1 1782 121
XM_021087051.1 XP_020942710.1 1852 121
XM_021087052.1 XP_020942711.1 450 121
XM_021087053.1 XP_020942712.1 423 121
XM_021087054.1 XP_020942713.1 245 121
XM_021087055.1 XP_020942714.1 1782 121
XM_021087056.1 XP_020942715.1 1782 121
XM_021087057.1 XP_020942716.1 1782 121
XM_021087058.1 XP_020942717.1 1782 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0