This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06863653_s1
See other RAI2 GE Assays ›
Gene Symbol:
RAI2
Gene Name:
retinoic acid induced 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 14246776 - 14633071 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06863653_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAI2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013985950.2 XP_013841404.1 1520 67
XM_021080586.1 XP_020936245.1 2203 67
XM_021080587.1 XP_020936246.1 2263 67
XM_021080588.1 XP_020936247.1 1963 67
XM_021080589.1 XP_020936248.1 4395 67
XM_021080590.1 XP_020936249.1 2532 67
XM_021080591.1 XP_020936250.1 2042 67
XM_021080592.1 XP_020936251.1 1768 67
XM_021080594.1 XP_020936253.1 1675 67
XM_021080595.1 XP_020936254.1 1618 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline