This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06866698_g1
See other ENO1 GE Assays ›
Gene Symbol:
ENO1
Gene Name:
enolase 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 69385879 - 69401664 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06866698_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ENO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021095278.1 XP_020950937.1 67
XM_021095279.1 XP_020950938.1 67
XM_021095280.1 XP_020950939.1 67
GenBank mRNA AK232866.1 - 6 - 7 562 67
AK234314.1 - 6 - 7 582 67
AK234575.1 - 6 - 7 531 67
AK235429.1 - 5 - 6 442 67
AK238490.1 - 6 - 7 560 67
AK238537.1 - 6 - 7 561 67
AK393738.1 - 6 - 7 634 67
AK395106.1 - 6 - 7 531 67
AK396321.1 - 6 - 7 586 67
AK398021.1 - 6 - 7 587 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline