This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06867221_m1
See other MSRA GE Assays ›
Gene Symbol:
MSRA
Gene Name:
methionine sulfoxide reductase A
Gene Aliases:
-
Chromosome Location:
Chr.14: 13334731 - 13718914 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06867221_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003359049.4 XP_003359097.3 729 105
XM_021072631.1 XP_020928290.1 1205 105
XM_021072633.1 XP_020928292.1 1382 105
XM_021072634.1 XP_020928293.1 1330 105
XM_021072635.1 XP_020928294.1 1301 105
XM_021072636.1 XP_020928295.1 977 105
XM_021072637.1 XP_020928296.1 2941 105
XM_021072638.1 XP_020928297.1 778 105
XM_021072639.1 XP_020928298.1 1094 105
XM_021072640.1 XP_020928299.1 2463 105
XM_021072641.1 XP_020928300.1 1019 105
XM_021072643.1 XP_020928302.1 898 105
GenBank mRNA AK233136.1 - 5 - 6 632 105
AK233412.1 - 6 - 7 735 105
AK397564.1 - 4 - 5 732 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0