This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06867963_m1
See other NEIL1 GE Assays ›
Gene Symbol:
NEIL1
Gene Name:
nei like DNA glycosylase 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 58261761 - 58269001 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06867963_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEIL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001924543.4 XP_001924578.1 881 73
XM_005666146.3 XP_005666203.1 1155 73
XM_013978062.2 XP_013833516.1 1085 73
XM_013978064.2 XP_013833518.1 744 73
XM_021099028.1 XP_020954687.1 1721 73
XM_021099029.1 XP_020954688.1 1922 73
XM_021099030.1 XP_020954689.1 1278 73
XM_021099031.1 XP_020954690.1 1943 73
XM_021099033.1 XP_020954692.1 2013 73
GenBank mRNA AK233426.1 - 4 - 5 693 73
AK233534.1 - 4 - 5 708 73
AK395005.1 - 4 - 5 727 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline