This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06867978_m1
See other HDHD3 GE Assays ›
Gene Symbol:
HDHD3
Gene Name:
haloacid dehalogenase like hydrolase domain containing 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 254006018 - 254009804 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06867978_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDHD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021067738.1 XP_020923397.1 80
XM_021067746.1 XP_020923405.1 80
GenBank mRNA AK233430.1 - 1 - 2 65 80
AK390850.1 - 1 - 2 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021067738.1, XM_021067746.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline