This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06868457_m1
See other PPP2R3B GE Assays ›
Gene Symbol:
PPP2R3B
Gene Name:
protein phosphatase 2 regulatory subunit B''beta
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 90843 - 172647 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06868457_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021081364.1 XP_020937023.1 1719 59
XM_021081365.1 XP_020937024.1 1178 59
XM_021081366.1 XP_020937025.1 1716 59
XM_021081367.1 XP_020937026.1 1174 59
XM_021081369.1 XP_020937028.1 841 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-rd95r:80/100.66.77.199:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline