This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06869861_m1
See other ADGRG5 GE Assays ›
Gene Symbol:
ADGRG5
Gene Name:
adhesion G protein-coupled receptor G5
Gene Aliases:
GPR114
Chromosome Location:
Chr.6: 19456550 - 19488667 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06869861_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRG5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013988531.2 XP_013843985.2 2140 102
XM_021093911.1 XP_020949570.1 2120 102
XM_021093912.1 XP_020949571.1 2008 102
XM_021093913.1 XP_020949572.1 1828 102
XM_021093914.1 XP_020949573.1 2028 102
XM_021093915.1 XP_020949574.1 2195 102
XM_021093916.1 XP_020949575.1 1688 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013988531.2, XM_021093914.1, XM_021093915.1, XM_021093916.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rzthw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0