This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06873998_m1
See other ST18 GE Assays ›
Gene Symbol:
ST18
Gene Name:
ST18 C2H2C-type zinc finger transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.4: 77707143 - 78002016 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06873998_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ST18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021089378.1 XP_020945037.1 1592 59
XM_021089379.1 XP_020945038.1 905 59
XM_021089380.1 XP_020945039.1 908 59
XM_021089381.1 XP_020945040.1 827 59
XM_021089382.1 XP_020945041.1 992 59
XM_021089383.1 XP_020945042.1 824 59
XM_021089384.1 XP_020945043.1 935 59
XM_021089385.1 XP_020945044.1 707 59
XM_021089386.1 XP_020945045.1 1476 59
XM_021089388.1 XP_020945047.1 711 59
XM_021089389.1 XP_020945048.1 789 59
XM_021089390.1 XP_020945049.1 792 59
XM_021089391.1 XP_020945050.1 876 59
XM_021089392.1 XP_020945051.1 819 59
XM_021089393.1 XP_020945052.1 781 59
XM_021089394.1 XP_020945053.1 664 59
XM_021089395.1 XP_020945054.1 1443 59
XM_021089396.1 XP_020945055.1 1533 59
XM_021089397.1 XP_020945056.1 5292 59
XM_021089398.1 XP_020945057.1 5289 59
XM_021089399.1 XP_020945058.1 10115 59
XM_021089400.1 XP_020945059.1 10193 59
XM_021089401.1 XP_020945060.1 10196 59
XM_021089402.1 XP_020945061.1 10279 59
XM_021089403.1 XP_020945062.1 10112 59
XM_021089404.1 XP_020945063.1 5370 59
XM_021089405.1 XP_020945064.1 5373 59
XM_021089406.1 XP_020945065.1 5456 59
XM_021089407.1 XP_020945066.1 5719 59
XM_021089408.1 XP_020945067.1 5246 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0