This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06874716_s1
See other ELAVL2 GE Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like RNA binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 198964070 - 199135815 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06874716_s1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021062750.1
XP_020918409.1
1676
93
XM_021062755.1
XP_020918414.1
1670
93
XM_021062763.1
XP_020918422.1
1637
93
XM_021062771.1
XP_020918430.1
1632
93
XM_021062774.1
XP_020918433.1
1419
93
XM_021062780.1
XP_020918439.1
1246
93
XM_021062783.1
XP_020918442.1
1240
93
XM_021062788.1
XP_020918447.1
1244
93
XM_021062792.1
XP_020918451.1
1207
93
XM_021062802.1
XP_020918461.1
1624
93
XM_021062813.1
XP_020918472.1
1201
93
XM_021062823.1
XP_020918482.1
1194
93
XM_021062834.1
XP_020918493.1
1179
93
XM_021062839.1
XP_020918498.1
1599
93
XM_021062849.1
XP_020918508.1
1176
93
XM_021062861.1
XP_020918520.1
1167
93
XM_021062867.1
XP_020918526.1
1152
93
XM_021062871.1
XP_020918530.1
1375
93
XM_021062872.1
XP_020918531.1
1595
93
XM_021062874.1
XP_020918533.1
1279
93
XM_021062878.1
XP_020918537.1
1173
93
XM_021062881.1
XP_020918540.1
1306
93
XM_021062889.1
XP_020918548.1
1505
93
XM_021062900.1
XP_020918559.1
1279
93
XM_021062904.1
XP_020918563.1
1105
93
GenBank mRNA
KC155257.1
-
1
122
93
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
3' Most
Detect Genomic DNA
Recommended Assay
Sus scrofa
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
translation initiation factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0