This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06874857_m1
See other ATXN1 GE Assays ›
Gene Symbol:
ATXN1
Gene Name:
ataxin 1
Gene Aliases:
SCA1
Chromosome Location:
Chr.7: 12299938 - 12730850 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06874857_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013977443.2 XP_013832897.2 462 63
XM_013977445.2 XP_013832899.2 401 63
XM_013977446.2 XP_013832900.2 287 63
XM_013977447.2 XP_013832901.2 450 63
XM_013977449.2 XP_013832903.2 2014 63
XM_021099771.1 XP_020955430.1 506 63
XM_021099774.1 XP_020955433.1 7989 63
XM_021099775.1 XP_020955434.1 1085 63
GenBank mRNA AK233602.1 - 3 - 4 427 63
AK239951.1 - 3 - 4 469 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013977443.2, XM_013977445.2, XM_013977446.2, XM_013977447.2, XM_013977449.2, XM_021099771.1, XM_021099774.1, XM_021099775.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline