This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06875253_m1
See other DHRS9 GE Assays ›
Gene Symbol:
DHRS9
Gene Name:
dehydrogenase/reductase 9
Gene Aliases:
-
Chromosome Location:
Chr.15: 75507538 - 75533942 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06875253_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003133463.4 XP_003133511.1 1022 67
XM_013982216.2 XP_013837670.1 928 67
XM_021075144.1 XP_020930803.1 811 67
XM_021075145.1 XP_020930804.1 525 67
GenBank mRNA AK233817.1 - 2 - 3 561 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline