This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06876005_m1
See other ISLR GE Assays ›
Gene Symbol:
ISLR
Gene Name:
immunoglobulin superfamily containing leucine rich repeat
Gene Aliases:
MPI/7
Chromosome Location:
Chr.7: 59349674 - 59352936 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06876005_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ISLR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001928174.4 XP_001928209.1 419 81
GenBank mRNA AK234251.1 - 1 - 2 412 81
AK397468.1 - 1 - 2 424 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001928174.4 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline