This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06876084_m1
See other RBPMS GE Assays ›
Gene Symbol:
RBPMS
Gene Name:
RNA binding protein, mRNA processing factor
Gene Aliases:
-
Chromosome Location:
Chr.15: 54538583 - 54725605 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06876084_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBPMS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003133393.4 XP_003133441.3 875 73
XM_021074863.1 XP_020930522.1 9350 73
XM_021074864.1 XP_020930523.1 9353 73
XM_021074865.1 XP_020930524.1 9352 73
XM_021074866.1 XP_020930525.1 9362 73
XM_021074867.1 XP_020930526.1 298 73
XM_021074868.1 XP_020930527.1 9342 73
XM_021074869.1 XP_020930528.1 9339 73
XM_021074870.1 XP_020930529.1 9342 73
XM_021074871.1 XP_020930530.1 9346 73
XM_021074872.1 XP_020930531.1 683 73
XM_021074873.1 XP_020930532.1 9359 73
XM_021074874.1 XP_020930533.1 9353 73
XM_021074875.1 XP_020930534.1 9355 73
GenBank mRNA AK234271.1 - 2 - 3 659 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline