This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06878258_m1
See other HAGH GE Assays ›
Gene Symbol:
HAGH
Gene Name:
hydroxyacylglutathione hydrolase
Gene Aliases:
-
Chromosome Location:
Chr.3: 40139099 - 40162921 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06878258_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAGH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021086824.1 XP_020942483.1 3351 72
XM_021086825.1 XP_020942484.1 3351 72
XM_021086826.1 XP_020942485.1 3351 72
XM_021086827.1 XP_020942486.1 3351 72
XM_021086828.1 XP_020942487.1 3351 72
XM_021086829.1 XP_020942488.1 527 72
XM_021086830.1 XP_020942489.1 468 72
XM_021086831.1 XP_020942490.1 481 72
GenBank mRNA AK235240.1 - 4 - 5 532 72
AK236278.1 - 4 - 5 613 72
AK345979.1 - 4 - 5 454 72
AK350460.1 - 4 - 5 489 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0