This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06878270_m1
See other EIF5A GE Assays ›
Gene Symbol:
EIF5A
Gene Name:
eukaryotic translation initiation factor 5A
Gene Aliases:
-
Chromosome Location:
Chr.12: 52659040 - 52664686 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06878270_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003131931.4 XP_003131979.1 807 71
XM_003131932.4 XP_003131980.1 639 71
XM_013987951.2 XP_013843405.1 769 71
XM_021067815.1 XP_020923474.1 1946 71
XM_021067817.1 XP_020923476.1 557 71
GenBank mRNA AK235248.1 - 5 - 6 727 71
AK349939.1 - 5 - 6 610 71
AK389352.1 - 5 - 6 625 71
AK394081.1 - 5 - 6 617 71
AK394780.1 - 5 - 6 721 71
AK395145.1 - 5 - 6 733 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0