This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06878899_g1
See other GLMP GE Assays ›
Gene Symbol:
GLMP
Gene Name:
glycosylated lysosomal membrane protein
Gene Aliases:
C4H1orf85
Chromosome Location:
Chr.4: 93758939 - 93763517 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06878899_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLMP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001927751.3 XP_001927786.1 840 90
XM_005663295.3 XP_005663352.1 582 90
GenBank mRNA AK235511.1 - 4 - 5 829 90
AK390938.1 - 4 - 5 829 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0