This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06879463_g1
See other CNBP GE Assays ›
Gene Symbol:
CNBP
Gene Name:
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases:
-
Chromosome Location:
Chr.13: 71634552 - 71707027 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06879463_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021069418.1 XP_020925077.1 588 69
XM_021069419.1 XP_020925078.1 461 69
XM_021069420.1 XP_020925079.1 869 69
XM_021069421.1 XP_020925080.1 976 69
XM_021069423.1 XP_020925082.1 571 69
XM_021069424.1 XP_020925083.1 566 69
XM_021069425.1 XP_020925084.1 566 69
XM_021069426.1 XP_020925085.1 583 69
XM_021069427.1 XP_020925086.1 638 69
XM_021069428.1 XP_020925087.1 1037 69
XM_021069429.1 XP_020925088.1 559 69
XM_021069430.1 XP_020925089.1 556 69
GenBank mRNA AB529452.1 - 2 - 3 299 69
AK232643.1 - 4 - 5 578 69
AK235107.1 - 4 - 5 525 69
AK238902.1 - 4 - 5 548 69
AK351774.1 - 4 - 5 456 69
AK391085.1 - 4 - 5 565 69
AK394712.1 - 4 - 5 523 69
AK396189.1 - 4 - 5 578 69
AK397295.1 - 4 - 5 567 69
AK399552.1 - 4 - 5 576 69
AK399983.1 - 4 - 5 562 69
AK400100.1 - 4 - 5 581 69
AY609917.1 - 4 - 5 560 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0