This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06880569_g1
See other NDUFA5 GE Assays ›
Gene Symbol:
NDUFA5
Gene Name:
NADH:ubiquinone oxidoreductase subunit A5
Gene Aliases:
-
Chromosome Location:
Chr.18: 23881872 - 23896699 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06880569_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003134727.3 XP_003134775.1 354 77
XM_013985659.2 XP_013841113.1 200 77
GenBank mRNA AK230784.1 - 4 - 5 277 77
AK343776.1 - 4 - 5 277 77
AK343866.1 - 4 - 5 288 77
AK343910.1 - 4 - 5 288 77
AK392617.1 - 4 - 5 298 77
AY610277.1 - 4 - 5 292 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0