Assay Details
| Gene Symbol: ZNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086192.1 | XP_020941851.1 | 142 | 95 | ||
| GenBank mRNA | AK230805.1 | - | 1 - 2 | 107 | 95 | |
| AK231115.1 | - | 1 - 2 | 107 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086192.1 transcripts.
Set Membership: |
|

ZNF3
Genomic Map