This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06880778_g1
See other NDUFB3 GE Assays ›
Gene Symbol:
NDUFB3
Gene Name:
NADH:ubiquinone oxidoreductase subunit B3
Gene Aliases:
-
Chromosome Location:
Chr.15: 104747835 - 104759137 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06880778_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFB3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003133578.4 XP_003133626.1 395 131
XM_005652448.3 XP_005652505.1 386 131
XM_005652449.3 XP_005652506.1 469 131
XM_005652450.3 XP_005652507.1 462 131
XM_013996972.2 XP_013852426.1 294 131
XM_021075847.1 XP_020931506.1 455 131
XM_021075848.1 XP_020931507.1 383 131
XM_021075849.1 XP_020931508.1 415 131
XM_021075850.1 XP_020931509.1 384 131
XM_021075851.1 XP_020931510.1 249 131
XM_021075852.1 XP_020931511.1 375 131
XM_021075853.1 XP_020931512.1 464 131
XM_021075854.1 XP_020931513.1 455 131
GenBank mRNA AK230845.1 - 3 - 4 273 131
AK399697.1 - 3 - 4 293 131
AY610245.1 - 3 - 4 250 131

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0