This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06883960_s1
See other ARHGAP30 GE Assays ›
Gene Symbol:
ARHGAP30
Gene Name:
Rho GTPase activating protein 30
Gene Aliases:
-
Chromosome Location:
Chr.4: 89375486 - 89394190 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06883960_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001928060.7 XP_001928095.5 4215 115
XM_013996846.2 XP_013852300.1 4044 115
XM_021089539.1 XP_020945198.1 4328 115
XM_021089540.1 XP_020945199.1 4191 115
XM_021089541.1 XP_020945200.1 4146 115
XM_021089542.1 XP_020945201.1 4050 115
XM_021089543.1 XP_020945202.1 4196 115
XM_021089544.1 XP_020945203.1 4314 115
XM_021089545.1 XP_020945204.1 4186 115
XM_021089546.1 XP_020945205.1 4002 115
GenBank mRNA AK237181.1 - 1 1722 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0