This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06884329_s1
See other CHST15 GE Assays ›
Gene Symbol:
CHST15
Gene Name:
carbohydrate sulfotransferase 15
Gene Aliases:
-
Chromosome Location:
Chr.14: 133460159 - 133544771 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06884329_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHST15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005671535.3 XP_005671592.1 4593 67
XM_005671536.3 XP_005671593.1 4707 67
XM_005671537.3 XP_005671594.1 4993 67
XM_005671538.3 XP_005671595.1 4630 67
XM_005671540.3 XP_005671597.1 5967 67
XM_005671541.3 XP_005671598.1 4568 67
XM_005671542.3 XP_005671599.1 4582 67
XM_005671543.3 XP_005671600.1 4509 67
XM_005671544.3 XP_005671601.1 4505 67
XM_013983832.2 XP_013839286.1 4741 67
XM_013983836.2 XP_013839290.1 4842 67
XM_013983837.2 XP_013839291.1 6525 67
XM_021073610.1 XP_020929269.1 4731 67
XM_021073611.1 XP_020929270.1 4545 67
XM_021073612.1 XP_020929271.1 4472 67
XM_021073613.1 XP_020929272.1 4539 67
XM_021073615.1 XP_020929274.1 4391 67
XM_021073616.1 XP_020929275.1 4396 67
GenBank mRNA AK237412.1 - 1 1522 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline