This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06884361_m1
See other NUDT13 GE Assays ›
Gene Symbol:
NUDT13
Gene Name:
nudix hydrolase 13
Gene Aliases:
-
Chromosome Location:
Chr.14: 75918056 - 75958187 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06884361_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005671122.3 XP_005671179.2 550 80
XM_013983336.2 XP_013838790.1 441 80
XM_021074323.1 XP_020929982.1 783 80
XM_021074324.1 XP_020929983.1 1193 80
XM_021074325.1 XP_020929984.1 1911 80
XM_021074326.1 XP_020929985.1 1047 80
XM_021074328.1 XP_020929987.1 976 80
XM_021074329.1 XP_020929988.1 869 80
XM_021074330.1 XP_020929989.1 785 80
XM_021074331.1 XP_020929990.1 958 80
XM_021074332.1 XP_020929991.1 917 80
XM_021074333.1 XP_020929992.1 921 80
XM_021074334.1 XP_020929993.1 829 80
XM_021074335.1 XP_020929994.1 690 80
XM_021074336.1 XP_020929995.1 777 80
XM_021074338.1 XP_020929997.1 597 80
XM_021074339.1 XP_020929998.1 1115 80
XM_021074340.1 XP_020929999.1 1008 80
XM_021074341.1 XP_020930000.1 642 80
XM_021074342.1 XP_020930001.1 729 80
XM_021074343.1 XP_020930002.1 637 80
XM_021074344.1 XP_020930003.1 490 80
GenBank mRNA AK237426.1 - 8 - 9 1088 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0