This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06885746_m1
See other DISP1 GE Assays ›
Gene Symbol:
DISP1
Gene Name:
dispatched RND transporter family member 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 11526353 - 11709888 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06885746_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DISP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021064348.1 XP_020920007.1 1781 62
XM_021064349.1 XP_020920008.1 698 62
XM_021064350.1 XP_020920009.1 785 62
XM_021064351.1 XP_020920010.1 851 62
XM_021064352.1 XP_020920011.1 3079 62
XM_021064353.1 XP_020920012.1 750 62
XM_021064355.1 XP_020920014.1 2856 62
XM_021064356.1 XP_020920015.1 623 62
XM_021064357.1 XP_020920016.1 436 62
XM_021064358.1 XP_020920017.1 388 62
XM_021064359.1 XP_020920018.1 320 62
XM_021064360.1 XP_020920019.1 336 62
XM_021064361.1 XP_020920020.1 765 62
XM_021064362.1 XP_020920021.1 684 62
XM_021064363.1 XP_020920022.1 1845 62
GenBank mRNA AK238379.1 - 4 - 5 753 62
AK347473.1 - 4 - 5 750 62
AK351163.1 - 3 - 4 636 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021064363.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0