This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06886078_m1
See other NFE2L2 GE Assays ›
Gene Symbol:
NFE2L2
Gene Name:
nuclear factor, erythroid 2 like 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 82967485 - 83146185 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06886078_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFE2L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003133500.6 XP_003133548.1 906 95
XM_003483682.4 XP_003483730.1 748 95
XM_005671981.3 XP_005672038.1 1087 95
XM_013984303.2 XP_013839757.1 997 95
XM_021075130.1 XP_020930789.1 1091 95
XM_021075131.1 XP_020930790.1 876 95
XM_021075132.1 XP_020930791.1 661 95
XM_021075133.1 XP_020930792.1 742 95
GenBank mRNA AK231013.1 - 4 - 5 586 95
AK236642.1 - 4 - 5 700 95
AK389768.1 - 4 - 5 730 95
AK395865.1 - 4 - 5 648 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline