Assay Details
Gene Symbol: TP53I11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122849.6 | XP_003122897.3 | 500 | 57 | ||
XM_013994384.2 | XP_013849838.1 | 603 | 57 | |||
XM_021083156.1 | XP_020938815.1 | 816 | 57 | |||
GenBank mRNA | AK231375.1 | - | 4 - 5 | 352 | 57 | |
AK235421.1 | - | 4 - 5 | 460 | 57 | ||
AK349207.1 | - | 4 - 5 | 419 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
