This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06887290_m1
See other EIF3K GE Assays ›
Gene Symbol:
EIF3K
Gene Name:
eukaryotic translation initiation factor 3 subunit K
Gene Aliases:
-
Chromosome Location:
Chr.6: 47476184 - 47488933 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06887290_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003127119.5 XP_003127167.2 252 67
XM_005664595.3 XP_005664652.1 248 67
GenBank mRNA AK231473.1 - 2 - 3 480 67
AK233726.1 - 2 - 3 226 67
AK239915.1 - 2 - 3 229 67
AK392372.1 - 2 - 3 231 67
AK399887.1 - 2 - 3 240 67
AY610350.1 - 2 - 3 183 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0