This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06889606_m1
See other UNG GE Assays ›
Gene Symbol:
UNG
Gene Name:
uracil DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.14: 41746498 - 41757841 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06889606_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003132925.6 XP_003132973.1 723 75
GenBank mRNA AK239783.1 - 5 - 6 695 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline