This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06890651_m1
See other PELI3 GE Assays ›
Gene Symbol:
PELI3
Gene Name:
pellino E3 ubiquitin protein ligase family member 3
Gene Aliases:
-
Chromosome Location:
Chr.2: 5950552 - 5959786 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06890651_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PELI3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003122491.5 XP_003122539.1 810 80
XM_005660658.3 XP_005660715.1 738 80
XM_013994053.2 XP_013849507.1 896 80
XM_021082795.1 XP_020938454.1 930 80
XM_021082796.1 XP_020938455.1 744 80
XM_021082797.1 XP_020938456.1 691 80
XM_021082798.1 XP_020938457.1 585 80
GenBank mRNA AK343243.1 - 6 - 7 806 80
AK389922.1 - 5 - 6 681 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5lprg:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0