This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06891979_gH
See other RPL34 GE Assays ›
Gene Symbol:
RPL34
Gene Name:
ribosomal protein L34
Gene Aliases:
-
Chromosome Location:
Chr.8: 113467779 - 113472058 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06891979_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL34
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005666942.3 XP_005666999.1 123 107
XM_005666943.3 XP_005667000.1 160 107
XM_013978964.2 XP_013834418.1 203 107
XM_013978965.1 XP_013834419.1 1619 107
GenBank mRNA AK344035.1 - 2 - 3 102 107
AK346105.1 - 2 - 3 1627 107
AK391712.1 - 2 - 3 102 107
AK399748.1 - 2 - 3 124 107
AY610479.1 - 1 - 2 73 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline