Assay Details
Gene Symbol: TNFRSF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005668283.3 | XP_005668340.1 | 972 | 52 | ||
XM_005668284.3 | XP_005668341.1 | 658 | 52 | |||
XM_021065467.1 | XP_020921126.1 | 734 | 52 | |||
XM_021065468.1 | XP_020921127.1 | 659 | 52 | |||
XM_021065469.1 | XP_020921128.1 | 674 | 52 | |||
XM_021065470.1 | XP_020921129.1 | 561 | 52 | |||
GenBank mRNA | AK349412.1 | - | 5 - 6 | 943 | 52 | |
AK390050.1 | - | 5 - 6 | 954 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
