This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06900030_m1
See other ESRRG GE Assays ›
Gene Symbol:
ESRRG
Gene Name:
estrogen related receptor gamma
Gene Aliases:
ESRRG1
Chromosome Location:
Chr.10: 6710615 - 7371419 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06900030_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESRRG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013980019.2 XP_013835473.1 672 60
XM_021064106.1 XP_020919765.1 8613 60
XM_021064107.1 XP_020919766.1 2503 60
XM_021064108.1 XP_020919767.1 872 60
XM_021064109.1 XP_020919768.1 891 60
XM_021064110.1 XP_020919769.1 1416 60
XM_021064111.1 XP_020919770.1 827 60
XM_021064112.1 XP_020919771.1 1408 60
XM_021064113.1 XP_020919772.1 2640 60
XM_021064114.1 XP_020919773.1 964 60
XM_021064115.1 XP_020919774.1 1098 60
XM_021064116.1 XP_020919775.1 2866 60
XM_021064117.1 XP_020919776.1 2732 60
XM_021064118.1 XP_020919777.1 1139 60
XM_021064119.1 XP_020919778.1 2807 60
XM_021064120.1 XP_020919779.1 910 60
XM_021064121.1 XP_020919780.1 2758 60
XM_021064122.1 XP_020919781.1 1006 60
XM_021064123.1 XP_020919782.1 952 60
XM_021064124.1 XP_020919783.1 1047 60
XM_021064125.1 XP_020919784.1 1108 60
XM_021064126.1 XP_020919785.1 1740 60
XM_021064127.1 XP_020919786.1 819 60
XM_021064128.1 XP_020919787.1 3238 60
XM_021064129.1 XP_020919788.1 2495 60
XM_021064131.1 XP_020919790.1 739 60
GenBank mRNA AK349992.1 - 4 - 5 844 60
KP717449.1 - 4 - 5 703 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013980019.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0