This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06900046_gH
See other PYCR1 GE Assays ›
Gene Symbol:
PYCR1
Gene Name:
pyrroline-5-carboxylate reductase 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 1060028 - 1064808 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06900046_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PYCR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021066408.1 XP_020922067.1 705 65
XM_021066409.1 XP_020922068.1 843 65
XM_021066410.1 XP_020922069.1 850 65
XM_021066412.1 XP_020922071.1 972 65
XM_021066413.1 XP_020922072.1 791 65
XM_021066414.1 XP_020922073.1 666 65
XM_021066415.1 XP_020922074.1 666 65
XM_021066416.1 XP_020922075.1 673 65
XM_021066417.1 XP_020922076.1 816 65
GenBank mRNA AK349999.1 - 5 - 6 616 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0