This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06903057_m1
See other PPIL2 GE Assays ›
Gene Symbol:
PPIL2
Gene Name:
peptidylprolyl isomerase like 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 50206807 - 50232845 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06903057_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001929539.5 XP_001929574.1 1290 57
XM_003132990.4 XP_003133038.1 1227 57
XM_005670915.3 XP_005670972.1 1290 57
XM_005670916.3 XP_005670973.1 1290 57
XM_013983168.2 XP_013838622.2 1288 57
XM_021072281.1 XP_020927940.1 1227 57
GenBank mRNA AK231904.1 - 15 - 16 1269 57
AK239286.1 - 14 - 15 1209 57
AK348438.1 - 15 - 16 1262 57
AK351932.1 - 15 - 16 1255 57
AK394781.1 - 15 - 16 1258 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline