This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06914647_g1
See other PGGHG GE Assays ›
Gene Symbol:
PGGHG
Gene Name:
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases:
ATHL1
Chromosome Location:
Chr.2: 99254 - 104898 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06914647_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGGHG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003354417.4 XP_003354465.2 1777 63
XM_005655041.3 XP_005655098.2 1845 63
XM_013987654.2 XP_013843108.2 1811 63
XM_021082530.1 XP_020938189.1 1858 63
XM_021082531.1 XP_020938190.1 1769 63
XM_021082532.1 XP_020938191.1 1822 63
XM_021082533.1 XP_020938192.1 1777 63
GenBank mRNA AK397680.1 - 4 - 5 467 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

glycosidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0