This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06915381_g1
See other BMF GE Assays ›
Gene Symbol:
BMF
Gene Name:
Bcl2 modifying factor
Gene Aliases:
-
Chromosome Location:
Chr.1: 131178021 - 131199668 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06915381_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BMF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021097829.1 XP_020953488.1 1153 61
XM_021097831.1 XP_020953490.1 864 61
XM_021097833.1 XP_020953492.1 430 61
XM_021097837.1 XP_020953496.1 1669 61
XM_021097840.1 XP_020953499.1 1337 61
XM_021097841.1 XP_020953500.1 700 61
XM_021097842.1 XP_020953501.1 1149 61
XM_021097844.1 XP_020953503.1 955 61
XM_021097846.1 XP_020953505.1 890 61
XM_021097847.1 XP_020953506.1 850 61
XM_021097848.1 XP_020953507.1 659 61
XM_021097851.1 XP_020953510.1 1111 61
XM_021097853.1 XP_020953512.1 1291 61
XM_021097854.1 XP_020953513.1 2764 61
XM_021097855.1 XP_020953514.1 655 61
XM_021097857.1 XP_020953516.1 606 61
GenBank mRNA AK398333.1 - 5 - 6 850 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0