Assay Details
| Gene Symbol: COX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128623.4 | XP_003128671.1 | 586 | 141 | ||
| GenBank mRNA | AY609587.1 | - | 3 - 4 | 593 | 141 | |
More Information
Set Membership: |
|

COX16
Genomic Map