This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06916317_m1
See other PPIL3 GE Assays ›
Gene Symbol:
PPIL3
Gene Name:
peptidylprolyl isomerase like 3
Gene Aliases:
-
Chromosome Location:
Chr.15: 104569138 - 104583752 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06916317_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001925925.4 XP_001925960.1 487 108
XM_001925981.5 XP_001926016.1 724 108
XM_005672067.3 XP_005672124.1 515 108
XM_005672068.3 XP_005672125.1 809 108
XM_005672069.3 XP_005672126.1 428 108
XM_013984381.2 XP_013839835.1 1296 108
XM_021074952.1 XP_020930611.1 1246 108
GenBank mRNA AY609616.1 - 5 - 6 417 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline