Assay Details
| Gene Symbol: LOC106510546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094414.1 | XP_020950073.1 | 767 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106510546| Gene Symbol: LOC106510546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094414.1 | XP_020950073.1 | 767 | 83 | ||
Set Membership: |
|