This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06920520_m1
See other CLYBL GE Assays ›
Gene Symbol:
CLYBL
Gene Name:
citramalyl-CoA lyase
Gene Aliases:
-
Chromosome Location:
Chr.11: 68507828 - 68768322 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06920520_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLYBL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021065855.1 XP_020921514.1 594 70
XM_021065857.1 XP_020921516.1 594 70
XM_021065858.1 XP_020921517.1 594 70
XM_021065859.1 XP_020921518.1 594 70
XM_021065860.1 XP_020921519.1 594 70
XM_021065861.1 XP_020921520.1 594 70
XM_021065862.1 XP_020921521.1 594 70
XM_021065863.1 XP_020921522.1 2018 70
GenBank mRNA AK232461.1 - 4 - 5 545 70
AK238571.1 - 4 - 5 553 70
AK398207.1 - 4 - 5 578 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline