This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06920688_m1
See other ALDOA GE Assays ›
Gene Symbol:
ALDOA
Gene Name:
aldolase, fructose-bisphosphate A
Gene Aliases:
-
Chromosome Location:
Chr.3: 18246036 - 18260167 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06920688_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021087995.1 XP_020943654.1 623 69
XM_021087996.1 XP_020943655.1 441 69
XM_021087997.1 XP_020943656.1 1164 69
XM_021087998.1 XP_020943657.1 377 69
XM_021088000.1 XP_020943659.1 649 69
XM_021088001.1 XP_020943660.1 245 69
XM_021088002.1 XP_020943661.1 1954 69
XM_021088004.1 XP_020943663.1 306 69
GenBank mRNA AK232549.1 - 2 - 3 264 69
AK347107.1 - 2 - 3 170 69
AK392744.1 - 2 - 3 231 69
AK399722.1 - 2 - 3 252 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0