This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06927695_m1
See other IGSF5 GE Assays ›
Gene Symbol:
IGSF5
Gene Name:
immunoglobulin superfamily member 5
Gene Aliases:
-
Chromosome Location:
Chr.13: 203325385 - 203372361 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06927695_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005657158.3 XP_005657215.1 1663 124
XM_005657159.3 XP_005657216.1 1191 124
XM_021070993.1 XP_020926652.1 1299 124
XM_021070994.1 XP_020926653.1 1202 124
XM_021070996.1 XP_020926655.1 1176 124
XM_021070997.1 XP_020926656.1 1158 124
XM_021070998.1 XP_020926657.1 1318 124
XM_021070999.1 XP_020926658.1 999 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0