This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06928004_g1
See other GCGR GE Assays ›
Gene Symbol:
GCGR
Gene Name:
glucagon receptor
Gene Aliases:
-
Chromosome Location:
Chr.12: 1149589 - 1157234 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06928004_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCGR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021066441.1 XP_020922100.1 4801 64
XM_021066442.1 XP_020922101.1 814 64
XM_021066443.1 XP_020922102.1 768 64
XM_021066444.1 XP_020922103.1 531 64
XM_021066445.1 XP_020922104.1 814 64
XM_021066446.1 XP_020922105.1 4692 64
XM_021066447.1 XP_020922106.1 4255 64
XM_021066448.1 XP_020922107.1 2796 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline