Assay Details
| Gene Symbol: ZNF518A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013983631.2 | XP_013839085.1 | 530 | 89 | ||
| XM_013983632.2 | XP_013839086.1 | 757 | 89 | |||
| XM_013983633.2 | XP_013839087.1 | 1844 | 89 | |||
| XM_013983634.2 | XP_013839088.1 | 482 | 89 | |||
| XM_013983635.2 | XP_013839089.1 | 803 | 89 | |||
| XM_013983636.2 | XP_013839090.1 | 709 | 89 | |||
| XM_013983637.2 | XP_013839091.1 | 305 | 89 | |||
| XM_013983638.2 | XP_013839092.1 | 947 | 89 | |||
| XM_013983640.2 | XP_013839094.1 | 396 | 89 | |||
| XM_021074255.1 | XP_020929914.1 | 1585 | 89 | |||
| XM_021074256.1 | XP_020929915.1 | 1797 | 89 | |||
| XM_021074257.1 | XP_020929916.1 | 352 | 89 | |||
| XM_021074258.1 | XP_020929917.1 | 305 | 89 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013983631.2, XM_013983632.2, XM_013983633.2, XM_013983634.2, XM_013983635.2, XM_013983636.2, XM_013983637.2, XM_013983638.2, XM_013983640.2, XM_021074255.1, XM_021074256.1, XM_021074257.1, XM_021074258.1 transcripts.
Set Membership: |
|

ZNF518A
Genomic Map