Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021076671.1 | XP_020932330.1 | 227 | 82 | ||
| XM_021076676.1 | XP_020932335.1 | 226 | 82 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021076671.1, XM_021076676.1 transcripts.
Set Membership: |
|

ZNF131
Genomic Map