This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06930996_m1
See other HELB GE Assays ›
Gene Symbol:
HELB
Gene Name:
DNA helicase B
Gene Aliases:
-
Chromosome Location:
Chr.5: 30645378 - 30696488 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06930996_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HELB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003126364.4 XP_003126412.1 888 75
XM_013997699.2 XP_013853153.1 345 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013997699.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

RNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0